Learn More
Medchemexpress LLC HY-130324 100mg Medchemexpress, Azido-PEG7-amine CAS:1333154-77-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130324100MG
Encompass_Preferred
Medchemexpress, HY-130324 100mg Azido-PEG7-amine CAS:1333154-77-0 Azido-PEG7-amine is a non-cleavable 7 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0203