Learn More
Medchemexpress LLC Azido-PEG7-amine | 1333154-77-0 | MFCD13185016 | >98.0% | 394.46 g/mol | C16H34N4O7 | 50 MG
SDP
Catalog No. 5000202258
Encompass_Preferred
Azido-PEG7-amine is a bifunctional polyethylene glycol (PEG) linker bearing an azide and a primary amine terminus. It is used as a click-chemistry building block and as a linker in antibody-drug conjugate (ADC) and PROTAC synthesis.
  • Bifunctional azide and primary amine termini suitable for conjugation.
  • PEG7 spacer provides a short, flexible linker (seven ethylene glycol units).
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted click reactions.
  • High reported purity, typically greater than 98%.
  • Supplied in small laboratory pack sizes for research use.
Catalog No. 50-002-02258 Supplier Medchemexpress LLC Supplier No. HY13032450MG
Azido-PEG7-amine is a bifunctional polyethylene glycol (PEG) linker bearing an azide and a primary amine terminus. It is used as a click-chemistry building block and as a linker in antibody-drug conjugate (ADC) and PROTAC synthesis.
  • Bifunctional azide and primary amine termini suitable for conjugation.
  • PEG7 spacer provides a short, flexible linker (seven ethylene glycol units).
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted click reactions.
  • High reported purity, typically greater than 98%.
  • Supplied in small laboratory pack sizes for research use.