Learn More
Medchemexpress LLC Azido-PEG7-azide | 225523-86-4 | 95.0% | C16H32N6O7 | 10MG
SDP
Catalog No. 5000260374
Encompass_Preferred
Azido-PEG7-azide is a bifunctional polyethylene glycol (PEG7) linker terminating in azide groups. It is used as a click-chemistry reagent and as a PROTAC linker to join molecules via azide-alkyne cycloaddition, providing a hydrophilic spacer that improves solubility and molecular flexibility for bioconjugation and linker synthesis.
  • Terminal azide groups suitable for CuAAC and SPAAC reactions.
  • Seven-unit PEG spacer increases solubility and reduces steric hindrance.
  • Useful for bioconjugation and PROTAC linker construction.
  • Offered in multiple small package sizes for research use.
  • Store dry and cool; in solution store at -80°C (long term) or -20°C (short term).
Catalog No. 50-002-60374 Supplier Medchemexpress LLC Supplier No. HY13339410MG
Azido-PEG7-azide is a bifunctional polyethylene glycol (PEG7) linker terminating in azide groups. It is used as a click-chemistry reagent and as a PROTAC linker to join molecules via azide-alkyne cycloaddition, providing a hydrophilic spacer that improves solubility and molecular flexibility for bioconjugation and linker synthesis.
  • Terminal azide groups suitable for CuAAC and SPAAC reactions.
  • Seven-unit PEG spacer increases solubility and reduces steric hindrance.
  • Useful for bioconjugation and PROTAC linker construction.
  • Offered in multiple small package sizes for research use.
  • Store dry and cool; in solution store at -80°C (long term) or -20°C (short term).