Learn More
Medchemexpress LLC Azido-PEG8-amine | 857891-82-8 | 99.9% | C18H38N4O8 | 50MG
SDP
Catalog No. 5000357244
Encompass_Preferred
Azido-PEG8-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne group molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-57244 Supplier Medchemexpress LLC Supplier No. HY14021650MG
Azido-PEG8-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne group molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions