Learn More
Medchemexpress LLC Azido-PEG8-azide | 361543-07-9 | 98.2% | 464.51 | 250 MG
SDP
Catalog No. 5000349371
Encompass_Preferred
Azido-PEG8-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Azide group that enables its participation in various cycloaddition reactions. This compound is suitable for both copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group for reaction versatility
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • For research use only
Catalog No. 50-003-49371 Supplier Medchemexpress LLC Supplier No. 250MGHY138706
Azido-PEG8-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent, featuring an Azide group that enables its participation in various cycloaddition reactions. This compound is suitable for both copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an Azide group for reaction versatility
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • For research use only