Learn More
Medchemexpress LLC HY-130742 100mg Medchemexpress, Azido-PEG8-Boc CAS:1623791-99-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130742100MG
Encompass_Preferred
Medchemexpress, HY-130742 100mg Azido-PEG8-Boc CAS:1623791-99-0 Azido-PEG8-Boc is a PEG- and Alkyl/ether-based PROTAC linker can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1250