Learn More
Medchemexpress LLC Azido-PEG8-hydrazide | 2353410-11-2 | 96.1% | C19H39N5O9 | 25 MG
SDP
Catalog No. 5000352659
Encompass_Preferred
Azido-PEG8-hydrazide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs, which are composed of two different ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions
  • Participates in SPAAC reactions
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-52659 Supplier Medchemexpress LLC Supplier No. 25MGHY140815
Azido-PEG8-hydrazide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with Alkyne groups. It can also engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs, which are composed of two different ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc reactions
  • Participates in SPAAC reactions
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins via the ubiquitin-proteasome system