Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG8-NHS ester | 1204834-00-3 | 98.0% | 564.6 g/mol | C23H40N4O12 | 25 MG
SDP

Catalog No. 5000202466
Encompass_Preferred

Azido-PEG8-NHS ester is an 8-unit polyethylene glycol (PEG) bifunctional linker bearing an azide on one terminus and an N-hydroxysuccinimide (NHS) ester on the other. It is used for bioconjugation: the azide enables azide-alkyne cycloaddition (copper-catalyzed or strain-promoted), while the NHS ester reacts with primary amines to form stable amide bonds.

  • Azide enables copper-catalyzed and strain-promoted click chemistry.
  • N-hydroxysuccinimide ester labels primary amines (for lysine and amine-modified substrates).
  • PEG8 spacer increases hydrophilicity and improves solubility in organic and mixed solvents.
  • Soluble in DMSO, dichloromethane, and DMF.
  • Commonly used as an ADC or PROTAC linker and general bioconjugation reagent.
  • Store at -20 °C to preserve stability.

Catalog No. 50-002-02466 Supplier Medchemexpress LLC Supplier No. HY13018425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG8-NHS ester is an 8-unit polyethylene glycol (PEG) bifunctional linker bearing an azide on one terminus and an N-hydroxysuccinimide (NHS) ester on the other. It is used for bioconjugation: the azide enables azide-alkyne cycloaddition (copper-catalyzed or strain-promoted), while the NHS ester reacts with primary amines to form stable amide bonds.

  • Azide enables copper-catalyzed and strain-promoted click chemistry.
  • N-hydroxysuccinimide ester labels primary amines (for lysine and amine-modified substrates).
  • PEG8 spacer increases hydrophilicity and improves solubility in organic and mixed solvents.
  • Soluble in DMSO, dichloromethane, and DMF.
  • Commonly used as an ADC or PROTAC linker and general bioconjugation reagent.
  • Store at -20 °C to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.