Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG8-NHS ester | 1204834-00-3 | 99.5% | 564.58 g/mol | C23H40N4O12 | 50 MG
SDP

Catalog No. 5000202260
Encompass_Preferred

Azido-PEG8-NHS ester is an azide-functional PEG8 N-hydroxysuccinimide (NHS) ester used as a cleavable linker and bioconjugation reagent for click chemistry and the synthesis of antibody-drug conjugates and PROTAC linkers. It is supplied as a colorless to light yellow liquid with high purity and is intended for research use.

  • Azide functionality enables copper-catalyzed and strain-promoted click reactions.
  • Peg8 spacer increases hydrophilicity and reduces steric hindrance.
  • NHS ester readily reacts with primary amines for bioconjugation.
  • High reported purity suitable for synthetic and bioconjugation workflows.
  • Storage stability: stable as a pure compound at -20°C for up to 3 years.
  • Solubility: soluble in DMSO (100 mg/mL); ultrasonic agitation may be required.

Catalog No. 50-002-02260 Supplier Medchemexpress LLC Supplier No. HY13018450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG8-NHS ester is an azide-functional PEG8 N-hydroxysuccinimide (NHS) ester used as a cleavable linker and bioconjugation reagent for click chemistry and the synthesis of antibody-drug conjugates and PROTAC linkers. It is supplied as a colorless to light yellow liquid with high purity and is intended for research use.

  • Azide functionality enables copper-catalyzed and strain-promoted click reactions.
  • Peg8 spacer increases hydrophilicity and reduces steric hindrance.
  • NHS ester readily reacts with primary amines for bioconjugation.
  • High reported purity suitable for synthetic and bioconjugation workflows.
  • Storage stability: stable as a pure compound at -20°C for up to 3 years.
  • Solubility: soluble in DMSO (100 mg/mL); ultrasonic agitation may be required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.