Learn More
Medchemexpress LLC Azido-PEG8-NHS ester | 1204834-00-3 | MFCD13184949 | 99.5% | 564.58 | C23H40N4O12 | 1 G
SDP
Catalog No. 5000202256
Encompass_Preferred
Azido-PEG8-NHS ester is an azide-functional polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester reactive group used for amine bioconjugation and click chemistry in ADC and PROTAC linker synthesis.
  • Enables NHS-amine coupling to primary amines.
  • Azide group enables copper-catalyzed and strain-promoted click chemistry.
  • PEG8 spacer increases solubility and reduces steric hindrance.
  • High reported purity for reliable conjugation performance.
  • Available in small laboratory pack suitable for research-scale work.
Catalog No. 50-002-02256 Supplier Medchemexpress LLC Supplier No. HY1301841G
Azido-PEG8-NHS ester is an azide-functional polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester reactive group used for amine bioconjugation and click chemistry in ADC and PROTAC linker synthesis.
  • Enables NHS-amine coupling to primary amines.
  • Azide group enables copper-catalyzed and strain-promoted click chemistry.
  • PEG8 spacer increases solubility and reduces steric hindrance.
  • High reported purity for reliable conjugation performance.
  • Available in small laboratory pack suitable for research-scale work.