Learn More
Broadpharm Azido-PEG8-triethoxysilane, 1g | Formula: C28H58N4O12Si | Purity: 98%
Catalog No. NC3662228
Encompass
Azido-PEG8-triethoxysilane is a short PEG linker featuring an azide, which is used in click chemistry with alkynes, and a triethoxysilane group, which may be used to retain a compound on silica surfaces such as those used in normal phase chromatography.
Catalog No. NC3662228 Supplier Broadpharm Supplier No. BP40349
Azido-PEG8-triethoxysilane is a short PEG linker featuring an azide, which is used in click chemistry with alkynes, and a triethoxysilane group, which may be used to retain a compound on silica surfaces such as those used in normal phase chromatography.