Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG9-acid | 1670249-37-2 | MFCD13184977 | 97.7% | 511.56 | C21H41N3O11 | 500 MG
SDP

Catalog No. 5000203914
Encompass_Preferred

Azido-PEG9-acid is a non-cleavable 9-unit polyethylene glycol (PEG) linker bearing an azide and a terminal carboxylic acid, used for conjugation in antibody-drug conjugate (ADC) and PROTAC synthesis. The azide enables click chemistry (CuAAC and SPAAC), while the carboxylic acid allows amide coupling to amines.

  • Non-cleavable 9-unit polyethylene glycol linker.
  • Contains an azide group for copper-catalyzed and strain-promoted click chemistry.
  • Terminal carboxylic acid suitable for amide coupling reactions.
  • Molecular weight about 511.56.
  • Purity approximately 97.7% (per manufacturer datasheet).
  • Available in 100 MG, 250 MG, 500 MG, 1 G, and 5 G pack sizes.
  • Appearance: colorless to light yellow liquid.
  • Storage: store pure material at -20°C for long-term stability.

Catalog No. 50-002-03914 Supplier Medchemexpress LLC Supplier No. HY130475500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG9-acid is a non-cleavable 9-unit polyethylene glycol (PEG) linker bearing an azide and a terminal carboxylic acid, used for conjugation in antibody-drug conjugate (ADC) and PROTAC synthesis. The azide enables click chemistry (CuAAC and SPAAC), while the carboxylic acid allows amide coupling to amines.

  • Non-cleavable 9-unit polyethylene glycol linker.
  • Contains an azide group for copper-catalyzed and strain-promoted click chemistry.
  • Terminal carboxylic acid suitable for amide coupling reactions.
  • Molecular weight about 511.56.
  • Purity approximately 97.7% (per manufacturer datasheet).
  • Available in 100 MG, 250 MG, 500 MG, 1 G, and 5 G pack sizes.
  • Appearance: colorless to light yellow liquid.
  • Storage: store pure material at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.