Learn More
Medchemexpress LLC Azido-PEG9-alcohol | 1984776-37-5 | 99.9% | C18H37N3O9 | 100 MG
SDP
Catalog No. 5000353798
Encompass_Preferred
Azido-PEG9-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating an Azide group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It is also capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53798 Supplier Medchemexpress LLC Supplier No. HY140800100MG
Azido-PEG9-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating an Azide group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It is also capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions