Learn More
Medchemexpress LLC Azido-PEG9-alcohol | 1984776-37-5 | 99.95% | C18H37N3O9 | 50 MG
SDP
Catalog No. 5000354970
Encompass_Preferred
Azido-PEG9-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group.
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Leverages the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
Catalog No. 50-003-54970 Supplier Medchemexpress LLC Supplier No. HY14080050MG
Azido-PEG9-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group.
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Leverages the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.