Learn More
Medchemexpress LLC Azido-PEG9-amine | 1207714-69-9 | 98.5% | 482.57 g/mol | C20H42N4O9 | 250 MG
SDP
Catalog No. 5000203827
Encompass_Preferred
Azido-PEG9-amine is an azide-terminated polyethylene glycol linker containing nine ethylene glycol units and a terminal primary amine, used as a click-chemistry reagent and as a non-cleavable PEG9 linker in bioconjugation applications.
  • Azide-terminated PEG linker with nine ethylene glycol units.
  • Terminal primary amine for further conjugation.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Suitable for antibody-drug conjugate (ADC) and proteolysis targeting chimera (PROTAC) synthesis and general bioconjugation.
  • CAS number 1207714-69-9; molecular weight 482.57 g/mol; formula C20H42N4O9; purity 98.5%.
  • Supplied as 250 mg quantity for laboratory-scale synthesis.
Catalog No. 50-002-03827 Supplier Medchemexpress LLC Supplier No. HY130169250MG
Azido-PEG9-amine is an azide-terminated polyethylene glycol linker containing nine ethylene glycol units and a terminal primary amine, used as a click-chemistry reagent and as a non-cleavable PEG9 linker in bioconjugation applications.
  • Azide-terminated PEG linker with nine ethylene glycol units.
  • Terminal primary amine for further conjugation.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Suitable for antibody-drug conjugate (ADC) and proteolysis targeting chimera (PROTAC) synthesis and general bioconjugation.
  • CAS number 1207714-69-9; molecular weight 482.57 g/mol; formula C20H42N4O9; purity 98.5%.
  • Supplied as 250 mg quantity for laboratory-scale synthesis.