Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG9-amine | 1207714-69-9 | 98.5% | 482.57 g/mol | C20H42N4O9 | 50 MG
SDP

Catalog No. 5000202255
Encompass_Preferred

Azido-PEG9-amine is a non-cleavable nine-unit polyethylene glycol (PEG) linker bearing an azide and a terminal amine. It is used for antibody-drug conjugate and PROTAC synthesis and enables bioconjugation via click chemistry (CuAAC and SPAAC).

  • Molecular formula: C20H42N4O9.
  • Molecular weight: 482.57 g/mol.
  • Cas number: 1207714-69-9.
  • Purity: 98.52%.
  • Available package sizes: 50 mg, 100 mg, 250 mg, 500 mg, 1 g.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted alkyne-azide cycloaddition for bioconjugation.

Catalog No. 50-002-02255 Supplier Medchemexpress LLC Supplier No. HY13016950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG9-amine is a non-cleavable nine-unit polyethylene glycol (PEG) linker bearing an azide and a terminal amine. It is used for antibody-drug conjugate and PROTAC synthesis and enables bioconjugation via click chemistry (CuAAC and SPAAC).

  • Molecular formula: C20H42N4O9.
  • Molecular weight: 482.57 g/mol.
  • Cas number: 1207714-69-9.
  • Purity: 98.52%.
  • Available package sizes: 50 mg, 100 mg, 250 mg, 500 mg, 1 g.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted alkyne-azide cycloaddition for bioconjugation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.