Learn More
Medchemexpress LLC Azido-PEG9-Boc | 1818294-43-7 | C25H49N3O11 | 100 MG
SDP
Catalog No. 5000353160
Encompass_Preferred
Azido-PEG9-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-53160 Supplier Medchemexpress LLC Supplier No. HY140780100MG
Azido-PEG9-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)