Learn More
Medchemexpress LLC Azido-PEG9-NHS ester | 2801772-87-0 | >98% | C25H44N4O13 | 50 MG
SDP
Catalog No. 5000355695
Encompass_Preferred
Azido-PEG9-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with compounds containing DBCO or BCN groups. For optimal stability, store in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and ignition sources.
  • Can be used in the synthesis of PROTACs
  • Features an Azide group for click chemistry applications
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Recommended storage at -20°C for up to 3 years in pure form
Catalog No. 50-003-55695 Supplier Medchemexpress LLC Supplier No. HY14075850MG
Azido-PEG9-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent features an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with compounds containing DBCO or BCN groups. For optimal stability, store in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and ignition sources.
  • Can be used in the synthesis of PROTACs
  • Features an Azide group for click chemistry applications
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Recommended storage at -20°C for up to 3 years in pure form