Learn More
Cayman Chemical AzIdo-ThalIdomIde 50mg
SDP
Catalog No. 502933320
Encompass_Preferred
A PROTAC building block; contains thalidomide, which is a ligand for the E3 ubiquitin ligase cereblon, and can be conjugated to alkynylated ligands for target proteins via click chemistry; has been used in the synthesis of a SirReal-based PROTAC for the degradation of Sirt2 in cells
Catalog No. 50-293-3320 Supplier Cayman Chemical Supplier No. 2614650 MG
A PROTAC building block; contains thalidomide, which is a ligand for the E3 ubiquitin ligase cereblon, and can be conjugated to alkynylated ligands for target proteins via click chemistry; has been used in the synthesis of a SirReal-based PROTAC for the degradation of Sirt2 in cells