Learn More
Medchemexpress LLC Azidoacetic Acid | 18523-48-3 | MFCD01320907 | C2H3N3O2 | 5 G
SDP
Catalog No. 5000374121
Encompass_Preferred
Azidoacetic Acid is a click chemistry reagent that contains an azide group. It can be utilized as a small molecule tool for the synthesis of PROTAC. This compound can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain alkyne groups, and can also undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Utilized as a small molecule tool for PROTAC synthesis
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-74121 Supplier Medchemexpress LLC Supplier No. HY1517195G
Add to Cart
Add to Cart
Azidoacetic Acid is a click chemistry reagent that contains an azide group. It can be utilized as a small molecule tool for the synthesis of PROTAC. This compound can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain alkyne groups, and can also undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Utilized as a small molecule tool for PROTAC synthesis
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)