Learn More
Medchemexpress LLC Azidoethyl-SS-ethylamine | 1807512-40-8 | 98.0% | 178.28 | C4H10N4S2 | 5 MG
SDP
Catalog No. 5000204166
Encompass_Preferred
Azidoethyl-SS-ethylamine is a cleavable disulfide antibody-drug conjugate (ADC) linker bearing an azide functional group for click chemistry. It is used to install azide handles for subsequent conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), and is supplied in small quantities for research use.
  • Cleavable disulfide linker suitable for ADC construction.
  • Azide functional group enables CuAAC and SPAAC conjugation.
  • High purity (98.0%) for research applications.
  • Low molecular weight (178.28 g/mol) to minimize steric effects.
  • Available in small research quantities (5 mg) for preparative work.
Catalog No. 50-002-04166 Supplier Medchemexpress LLC Supplier No. HY1401045MG
Azidoethyl-SS-ethylamine is a cleavable disulfide antibody-drug conjugate (ADC) linker bearing an azide functional group for click chemistry. It is used to install azide handles for subsequent conjugation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), and is supplied in small quantities for research use.
  • Cleavable disulfide linker suitable for ADC construction.
  • Azide functional group enables CuAAC and SPAAC conjugation.
  • High purity (98.0%) for research applications.
  • Low molecular weight (178.28 g/mol) to minimize steric effects.
  • Available in small research quantities (5 mg) for preparative work.