Learn More
Medchemexpress LLC Azidoethyl-SS-ethylamine | 1807512-40-8 | 98.0% | 178.28 g/mol | C4H10N4S2 | 10 MG
SDP
Catalog No. 5000203947
Encompass_Preferred
Azidoethyl-SS-ethylamine is a cleavable antibody-drug conjugate (ADC) linker containing an azide functional group used for click chemistry-based bioconjugation. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), facilitating linker installation in ADC synthesis and other conjugation workflows.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Supports CuAAC and SPAAC click chemistry reactions.
  • High purity (98.0%) and defined molecular weight (178.28 g/mol).
  • Small pack sizes suitable for research-scale work (e.g., 10 mg).
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only.
Catalog No. 50-002-03947 Supplier Medchemexpress LLC Supplier No. HY14010410MG
Azidoethyl-SS-ethylamine is a cleavable antibody-drug conjugate (ADC) linker containing an azide functional group used for click chemistry-based bioconjugation. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), facilitating linker installation in ADC synthesis and other conjugation workflows.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Supports CuAAC and SPAAC click chemistry reactions.
  • High purity (98.0%) and defined molecular weight (178.28 g/mol).
  • Small pack sizes suitable for research-scale work (e.g., 10 mg).
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only.