Learn More
Medchemexpress LLC Azidoethyl-SS-ethylamine | 1807512-40-8 | MFCD28976704 | 98.0% | 178.28 g/mol | C4H10N4S2 | 50 MG
SDP
Catalog No. 5000203864
Encompass_Preferred
Azidoethyl-SS-ethylamine is a cleavable antibody-drug conjugate linker and click-chemistry reagent containing an azide and a disulfide moiety. It is used to introduce an azidoethyl-disulfide handle for conjugation by copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC) in bioconjugation and ADC synthesis.
  • Cleavable disulfide linker for controlled payload release
  • Reactive azide group for CuAAC and SPAAC conjugation
  • Compatible with antibody-drug conjugate synthesis and bioconjugation workflows
  • Stable in pure form when stored at -20°C
Catalog No. 50-002-03864 Supplier Medchemexpress LLC Supplier No. HY14010450MG
Azidoethyl-SS-ethylamine is a cleavable antibody-drug conjugate linker and click-chemistry reagent containing an azide and a disulfide moiety. It is used to introduce an azidoethyl-disulfide handle for conjugation by copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC) in bioconjugation and ADC synthesis.
  • Cleavable disulfide linker for controlled payload release
  • Reactive azide group for CuAAC and SPAAC conjugation
  • Compatible with antibody-drug conjugate synthesis and bioconjugation workflows
  • Stable in pure form when stored at -20°C