Learn More
Medchemexpress LLC Azidoethyl-ss-ethylamine | 1807512-40-8 | MFCD32066832 | 98.0% | 178.28 g·mol-1 | C4H10N4S2 | 25 MG
SDP
Catalog No. 5000204246
Encompass_Preferred
Azidoethyl-ss-ethylamine is a cleavable antibody-drug conjugate (ADC) linker and click-chemistry reagent that contains an azide functional group used for copper-catalyzed and strain-promoted azide-alkyne cycloaddition bioconjugation reactions.
  • Azide functional group for click chemistry conjugation.
  • Cleavable disulfide linker for controlled release in bioconjugates.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) reagents.
  • Provided as a small, easy-to-handle solid for synthetic applications.
Catalog No. 50-002-04246 Supplier Medchemexpress LLC Supplier No. HY14010425MG
Azidoethyl-ss-ethylamine is a cleavable antibody-drug conjugate (ADC) linker and click-chemistry reagent that contains an azide functional group used for copper-catalyzed and strain-promoted azide-alkyne cycloaddition bioconjugation reactions.
  • Azide functional group for click chemistry conjugation.
  • Cleavable disulfide linker for controlled release in bioconjugates.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) reagents.
  • Provided as a small, easy-to-handle solid for synthetic applications.