Learn More
Medchemexpress LLC Azidoethyl-SS-ethylazide | 352305-38-5 | 98.8% | 204.28 | C4H8N6S2 | 25 MG
SDP
Catalog No. 5000202587
Encompass_Preferred
Azidoethyl-SS-ethylazide is a cleavable ADC linker and click-chemistry reagent that contains an azide group for conjugation. It is used to introduce an azide handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted alkyne-azide cycloaddition (SPAAC) during bioconjugation and antibody-drug conjugate synthesis.
  • cleavable ADC linker for bioconjugation
  • contains an azide functional group
  • compatible with CuAAC and SPAAC click reactions
  • used to introduce azide handles for downstream conjugation
  • available in small milligram pack sizes suitable for research
Catalog No. 50-002-02587 Supplier Medchemexpress LLC Supplier No. HY14010525MG
Azidoethyl-SS-ethylazide is a cleavable ADC linker and click-chemistry reagent that contains an azide group for conjugation. It is used to introduce an azide handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted alkyne-azide cycloaddition (SPAAC) during bioconjugation and antibody-drug conjugate synthesis.
  • cleavable ADC linker for bioconjugation
  • contains an azide functional group
  • compatible with CuAAC and SPAAC click reactions
  • used to introduce azide handles for downstream conjugation
  • available in small milligram pack sizes suitable for research