Learn More
Medchemexpress LLC Azidoethyl-ss-ethylazide | 352305-38-5 | MFCD24458414 | 96.0% | 204.28 g/mol | C4H8N6S2 | 50 MG
SDP
Catalog No. 5000202589
Encompass_Preferred
Azidoethyl-ss-ethylazide is a disulfide-cleavable azide linker used in bioconjugation and antibody-drug conjugate (ADC) synthesis. It contains an azide functional group for click chemistry and a disulfide bond for reductive cleavage, and is supplied for research use.
  • Cleavable disulfide linker suitable for ADC construction and conjugation workflows.
  • Azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click reactions.
  • Small-molecule linker with molecular weight about 204 g/mol and formula C4H8N6S2.
  • Store as recommended for long-term stability: pure material at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; consult supplier safety documentation for handling and disposal.
Catalog No. 50-002-02589 Supplier Medchemexpress LLC Supplier No. HY14010550MG
Azidoethyl-ss-ethylazide is a disulfide-cleavable azide linker used in bioconjugation and antibody-drug conjugate (ADC) synthesis. It contains an azide functional group for click chemistry and a disulfide bond for reductive cleavage, and is supplied for research use.
  • Cleavable disulfide linker suitable for ADC construction and conjugation workflows.
  • Azide functional group enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click reactions.
  • Small-molecule linker with molecular weight about 204 g/mol and formula C4H8N6S2.
  • Store as recommended for long-term stability: pure material at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; consult supplier safety documentation for handling and disposal.