Learn More
Medchemexpress LLC Azidoethyl-SS-PEG2-Boc | 2144777-83-1 | 351.49 | 100 MG
SDP
Catalog No. 5000349207
Encompass_Preferred
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs
Catalog No. 50-003-49207 Supplier Medchemexpress LLC Supplier No. HY140107100MG
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs