Learn More
Medchemexpress LLC Azidoethyl-SS-PEG2-Boc | 2144777-83-1 | 351.49 | 25 MG
SDP
Catalog No. 5000351701
Encompass_Preferred
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-51701 Supplier Medchemexpress LLC Supplier No. 25MGHY140107
Add to Cart
Add to Cart
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins