Learn More
Medchemexpress LLC Azidoethyl-SS-PEG2-Boc | 2144777-83-1 | 351.49 | 250 MG
SDP
Catalog No. 5000348259
Encompass_Preferred
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-48259 Supplier Medchemexpress LLC Supplier No. 250MGHY140107
Azidoethyl-SS-PEG2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins