Learn More
Cayman Chemical Azidoindolene 1, Purity: ≥98%, CAS: 1364933-69-6, 10 mg
SDP
Catalog No. 502966018
Encompass_Preferred
A synthetic CB structurally similar to UR-144, XLR11, and a number of other synthetic CBs with a hydrazide linking the tetramethylcyclopropyl group to the aminoalkylindole group; intended for forensic and research applications
Catalog No. 50-296-6018 Supplier Cayman Chemical Supplier No. 1708310 MG
A synthetic CB structurally similar to UR-144, XLR11, and a number of other synthetic CBs with a hydrazide linking the tetramethylcyclopropyl group to the aminoalkylindole group; intended for forensic and research applications