Learn More
Apexbio Technology LLC Azilsartan 147403-03-0 10mg
SDP
Catalog No. 502240780
Encompass_Preferred
Azilsartan (CAS 147403-03-0) is a potent small-molecule angiotensin II type 1 (AT1) receptor antagonist It competitively inhibits the binding of angiotensin II to the AT1 receptor exhibiting an IC50 of 2 6 nM By blocking this receptor azilsartan modulates the renin-angiotensin system which is critical in the regulation of blood pressure and cardiovascular function Azilsartan is widely utilized in research to investigate AT1 receptor signaling pathways and to explore mechanisms underlying hypertension and related cardiovascular disorders
This item is not returnable. View return policy
Catalog No. 50-224-0780 Supplier Apexbio Technology LLC Supplier No. B221010
Azilsartan (CAS 147403-03-0) is a potent small-molecule angiotensin II type 1 (AT1) receptor antagonist It competitively inhibits the binding of angiotensin II to the AT1 receptor exhibiting an IC50 of 2 6 nM By blocking this receptor azilsartan modulates the renin-angiotensin system which is critical in the regulation of blood pressure and cardiovascular function Azilsartan is widely utilized in research to investigate AT1 receptor signaling pathways and to explore mechanisms underlying hypertension and related cardiovascular disorders