Learn More
TARGETMOL CHEMICALS INC Azilsartan 200MG
SDP
Catalog No. 502880625
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Azilsartan (TAK-536) is an Angiotensin 2 Receptor Blocker that acts as an Angiotensin 2 Type 1 Receptor Antagonist, leading to a physiologic effect of decreased blood pressure. Purity 99.87%
Catalog No. 50-288-0625 Supplier TARGETMOL CHEMICALS INC Supplier No. T1057200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Azilsartan (TAK-536) is an Angiotensin 2 Receptor Blocker that acts as an Angiotensin 2 Type 1 Receptor Antagonist, leading to a physiologic effect of decreased blood pressure. Purity 99.87%