Learn More
Medchemexpress LLC Azilsartan-d5 | 1346599-45-8 | 99.4% | 461.48 | 5 MG
SDP
Catalog No. 5000361012
Encompass_Preferred
Azilsartan-d5 is the deuterium labeled Azilsartan (TAK-536), which is a specific and potent angiotensin II type 1 receptor antagonist. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-61012 Supplier Medchemexpress LLC Supplier No. HY14914S5MG
Azilsartan-d5 is the deuterium labeled Azilsartan (TAK-536), which is a specific and potent angiotensin II type 1 receptor antagonist. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.