Learn More
Medchemexpress LLC Azilsartan-d5 (TAK-536-d5) | 1346599-45-8 | 99.4% | 461.48 | 1 ML
SDP
Catalog No. 5000360888
Encompass_Preferred
Azilsartan-d5 is the deuterium-labeled Azilsartan (TAK-536), a specific and potent angiotensin II type 1 receptor antagonist. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled Azilsartan
  • Specific and potent angiotensin II type 1 receptor antagonist
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60888 Supplier Medchemexpress LLC Supplier No. HY14914S10MM/1ML
Azilsartan-d5 is the deuterium-labeled Azilsartan (TAK-536), a specific and potent angiotensin II type 1 receptor antagonist. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium-labeled Azilsartan
  • Specific and potent angiotensin II type 1 receptor antagonist
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS