Learn More
Apexbio Technology LLC Azilsartan Medoxomil 863031-21-4 250mg
SDP
Catalog No. 5000090448
Encompass_Preferred
Azilsartan Medoxomil (CAS 863031-21-4) is an orally active prodrug that is converted in vivo to azilsartan a selective antagonist of the angiotensin II type 1 (AT1) receptor By inhibiting AT1 receptor signaling it interrupts the renin-angiotensin-aldosterone system thereby modulating vascular resistance and blood pressure In biochemical assays azilsartan medoxomil demonstrates potent antagonistic activity with an IC50 of 0 62 nM This compound is widely utilized in research focused on cardiovascular disease mechanisms and the pharmacological modulation of hypertension
Catalog No. 50-000-90448 Supplier Apexbio Technology LLC Supplier No. B2218250
Azilsartan Medoxomil (CAS 863031-21-4) is an orally active prodrug that is converted in vivo to azilsartan a selective antagonist of the angiotensin II type 1 (AT1) receptor By inhibiting AT1 receptor signaling it interrupts the renin-angiotensin-aldosterone system thereby modulating vascular resistance and blood pressure In biochemical assays azilsartan medoxomil demonstrates potent antagonistic activity with an IC50 of 0 62 nM This compound is widely utilized in research focused on cardiovascular disease mechanisms and the pharmacological modulation of hypertension