Learn More
Medchemexpress LLC Azimilide | 149908-53-2 | 99.0% | 457.95 | 5 MG
SDP
Catalog No. 5000387847
Encompass_Preferred
Azimilide is a class III antiarrhythmic agent that blocks potassium channels in the heart. It is a dual blocker of IKs and IKr potassium channels. It also blocks HERG channels, and inhibits L-type calcium current (ICa) and sodium current (INa). It can be used for the study of atrial fibrillation and ventricular fibrillation.
  • Class III antiarrhythmic agent
  • Dual blocker of IKs and IKr potassium channels
  • Blocks HERG channels
  • Inhibits L-type calcium current (ICa)
  • Inhibits sodium current (INa)
  • Can be used for the study of atrial fibrillation and ventricular fibrillation
  • Significantly reduces the occurrence of ventricular tachycardia (VT) induced by electrical stimulation (PES) in dogs
  • Prolongs the effective refractory period of the ventricle and the QTc interval
  • Provides significant protection against ischemia-induced ventricular fibrillation and improves survival rates
Catalog No. 50-003-87847 Supplier Medchemexpress LLC Supplier No. HY186005MG
Azimilide is a class III antiarrhythmic agent that blocks potassium channels in the heart. It is a dual blocker of IKs and IKr potassium channels. It also blocks HERG channels, and inhibits L-type calcium current (ICa) and sodium current (INa). It can be used for the study of atrial fibrillation and ventricular fibrillation.
  • Class III antiarrhythmic agent
  • Dual blocker of IKs and IKr potassium channels
  • Blocks HERG channels
  • Inhibits L-type calcium current (ICa)
  • Inhibits sodium current (INa)
  • Can be used for the study of atrial fibrillation and ventricular fibrillation
  • Significantly reduces the occurrence of ventricular tachycardia (VT) induced by electrical stimulation (PES) in dogs
  • Prolongs the effective refractory period of the ventricle and the QTc interval
  • Provides significant protection against ischemia-induced ventricular fibrillation and improves survival rates