Learn More
Medchemexpress LLC Azimilide | 149908-53-2 | MFCD00867099 | 99.0% | 457.95 | C23H28ClN5O3 | 10 MG
SDP
Catalog No. 5000295064
Encompass_Preferred
Azimilide is a class III antiarrhythmic agent used in research to block cardiac delayed rectifier potassium channels (IKs and IKr). It is commonly used in electrophysiology and arrhythmia studies to probe ion channel function and pharmacological effects.
  • Class III potassium channel blocker targeting IKs and IKr.
  • Also inhibits L-type calcium and sodium currents at higher concentrations.
  • Suitable for in vitro electrophysiology and arrhythmia models.
  • High purity grade for research use.
  • Free base and dihydrochloride salt forms available; follow SDS storage guidance.
Catalog No. 50-002-95064 Supplier Medchemexpress LLC Supplier No. HY1860010MG
Azimilide is a class III antiarrhythmic agent used in research to block cardiac delayed rectifier potassium channels (IKs and IKr). It is commonly used in electrophysiology and arrhythmia studies to probe ion channel function and pharmacological effects.
  • Class III potassium channel blocker targeting IKs and IKr.
  • Also inhibits L-type calcium and sodium currents at higher concentrations.
  • Suitable for in vitro electrophysiology and arrhythmia models.
  • High purity grade for research use.
  • Free base and dihydrochloride salt forms available; follow SDS storage guidance.