Learn More
Apexbio Technology LLC Azimilide 149908-53-2 5mg
SDP
Catalog No. 5000063891
Encompass_Preferred
Azimilide (CAS 149908-53-2) is a small molecule under investigation for potential anti-arrhythmic activity classified pharmacologically as a class III anti-arrhythmic agent Its mechanism involves inhibition of cardiac delayed rectifier potassium channels specifically targeting both rapid (IKr) and slow (IKs) components By modulating these currents azimilide prolongs cardiac repolarization influencing cardiac rhythm Although not approved for therapeutic use azimilide is currently undergoing clinical evaluation in the United States and serves as a tool compound in arrhythmia-related biomedical research
Catalog No. 50-000-63891 Supplier Apexbio Technology LLC Supplier No. B48495
Azimilide (CAS 149908-53-2) is a small molecule under investigation for potential anti-arrhythmic activity classified pharmacologically as a class III anti-arrhythmic agent Its mechanism involves inhibition of cardiac delayed rectifier potassium channels specifically targeting both rapid (IKr) and slow (IKs) components By modulating these currents azimilide prolongs cardiac repolarization influencing cardiac rhythm Although not approved for therapeutic use azimilide is currently undergoing clinical evaluation in the United States and serves as a tool compound in arrhythmia-related biomedical research