Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azimilide dihydrochloride | 149888-94-8 | 98.2% | 530.88 | 1 ML
SDP

Catalog No. 5000388003
Encompass_Preferred

Azimilide dihydrochloride is a class III antiarrhythmic agent that functions by blocking potassium channels in the heart. It is a dual blocker of IKs (IC50 = 2.6 μM) and IKr (IC50 = 1 μM), and also inhibits HERG channels, L-type calcium current (ICa), and sodium current (INa). This compound is used for the study of atrial fibrillation and ventricular fibrillation.

  • Class III antiarrhythmic agent
  • Blocks potassium channels in the heart
  • Dual blocker of IKs and IKr
  • Inhibits HERG channel
  • Inhibits L-type calcium current (ICa) and sodium current (INa)
  • Can be used for the study of atrial fibrillation and ventricular fibrillation
  • Purity: 98.17%

Catalog No. 50-003-88003 Supplier Medchemexpress LLC Supplier No. HY18600A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azimilide dihydrochloride is a class III antiarrhythmic agent that functions by blocking potassium channels in the heart. It is a dual blocker of IKs (IC50 = 2.6 μM) and IKr (IC50 = 1 μM), and also inhibits HERG channels, L-type calcium current (ICa), and sodium current (INa). This compound is used for the study of atrial fibrillation and ventricular fibrillation.

  • Class III antiarrhythmic agent
  • Blocks potassium channels in the heart
  • Dual blocker of IKs and IKr
  • Inhibits HERG channel
  • Inhibits L-type calcium current (ICa) and sodium current (INa)
  • Can be used for the study of atrial fibrillation and ventricular fibrillation
  • Purity: 98.17%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.