Learn More
Medchemexpress LLC Azimilide dihydrochloride | 149888-94-8 | MFCD00918091 | >97.0% | 530.88 g/mol | C23H30Cl3N5O3 | 10 MG
SDP
Catalog No. 5000294882
Encompass_Preferred
Azimilide dihydrochloride is the dihydrochloride salt of azimilide, a class III antiarrhythmic research compound that blocks cardiac potassium currents. It is used in vitro to study cardiac electrophysiology, ion channel pharmacology, and mechanisms of arrhythmia in cellular and tissue models.
  • Dual IKs and IKr (hERG) blocker useful for cardiac electrophysiology studies.
  • Characterized IC50 values enable comparative ion channel pharmacology.
  • Offered as solid and as a ready-to-use 10 mM solution for assay flexibility.
  • Stable dihydrochloride salt with defined molecular weight and formula for reproducible dosing.
  • Suitable for in vitro investigations of atrial and ventricular fibrillation mechanisms.
Catalog No. 50-002-94882 Supplier Medchemexpress LLC Supplier No. HY18600A10MG
Azimilide dihydrochloride is the dihydrochloride salt of azimilide, a class III antiarrhythmic research compound that blocks cardiac potassium currents. It is used in vitro to study cardiac electrophysiology, ion channel pharmacology, and mechanisms of arrhythmia in cellular and tissue models.
  • Dual IKs and IKr (hERG) blocker useful for cardiac electrophysiology studies.
  • Characterized IC50 values enable comparative ion channel pharmacology.
  • Offered as solid and as a ready-to-use 10 mM solution for assay flexibility.
  • Stable dihydrochloride salt with defined molecular weight and formula for reproducible dosing.
  • Suitable for in vitro investigations of atrial and ventricular fibrillation mechanisms.