Learn More
Crescent Chemical Co Inc AZINOBIS(3-ETHYLBENZTHIAZOLINE
SDP
Catalog No. 50247329
Encompass_Preferred
Azinobis(3-ethylbenzthiazoline-6-sulfonic acid-2NH4-salt cryst. analytical grade Chromogen for peroxidase in enzyme-linked immunoassay (ELISA) (1). For laccase and angiotensin I-converting enzyme assay (2). Size - 1G Storage Conditions - +15 °C TO +30 °C Catalog Number - 14364.01 CAS N
Catalog No. 50-247-329 Supplier Crescent Chemical Co Inc Supplier No. SE14364.01
Azinobis(3-ethylbenzthiazoline-6-sulfonic acid-2NH4-salt cryst. analytical grade Chromogen for peroxidase in enzyme-linked immunoassay (ELISA) (1). For laccase and angiotensin I-converting enzyme assay (2). Size - 1G Storage Conditions - +15 °C TO +30 °C Catalog Number - 14364.01 CAS N