Learn More
Cayman Chemical Azithromycin-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2750534-82-6, 1 mg
SDP
Catalog No. 502989422
Encompass_Preferred
An internal standard for the quantification of azithromycin by GC- or LC-MS
Catalog No. 50-298-9422 Supplier Cayman Chemical Supplier No. 306531 MG
An internal standard for the quantification of azithromycin by GC- or LC-MS