Learn More
Medchemexpress LLC Azithromycin-13C,d3 | 2750534-82-6 | 99.9% | 753.00 | 500 UG
SDP
Catalog No. 5000386281
Encompass_Preferred
Azithromycin-13C,d3 is the deuterium and 13C labeled Azithromycin. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86281 Supplier Medchemexpress LLC Supplier No. HY17506S1500UG
Azithromycin-13C,d3 is the deuterium and 13C labeled Azithromycin. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.