Learn More
Cayman Chemical AzIthromycIn-13C-d3 500ug
SDP
Catalog No. 502988046
Encompass_Preferred
An internal standard for the quantification of azithromycin by GC- or LC-MS
Catalog No. 50-298-8046 Supplier Cayman Chemical Supplier No. 30653500 UG
An internal standard for the quantification of azithromycin by GC- or LC-MS