Learn More
Selleck Chemical LLC Azithromycin S1835-200mg
SDP
Catalog No. 503110922
Encompass_Preferred
Azithromycin is an antibiotic by inhibiting protein synthesis used for the treatment of bacterial infections
Catalog No. 50-311-0922 Supplier Selleck Chemical LLC Supplier No. S1835200MG
Azithromycin is an antibiotic by inhibiting protein synthesis used for the treatment of bacterial infections