Learn More
Apexbio Technology LLC Azithromycin 83905-01-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090845
Encompass_Preferred
Azithromycin (CAS 83905-01-5) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to inhibit microbial protein synthesis thereby interfering with peptide translation Azithromycin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies azithromycin demonstrates antiparasitic effects by inhibiting parasite propagation with an IC50 value of approximately 8 4 M Based on these pharmacological properties azithromycin holds research potential in investigating protein synthesis mechanisms and studying antimicrobial and antiparasitic drug resistance processes
Catalog No. 50-000-90845 Supplier Apexbio Technology LLC Supplier No. B13985.1
Azithromycin (CAS 83905-01-5) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to inhibit microbial protein synthesis thereby interfering with peptide translation Azithromycin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies azithromycin demonstrates antiparasitic effects by inhibiting parasite propagation with an IC50 value of approximately 8 4 M Based on these pharmacological properties azithromycin holds research potential in investigating protein synthesis mechanisms and studying antimicrobial and antiparasitic drug resistance processes