Learn More
Apexbio Technology LLC Azithromycin 83905-01-5 25mg
SDP
Catalog No. 5000089794
Encompass_Preferred
Azithromycin (CAS 83905-01-5) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to inhibit microbial protein synthesis thereby interfering with peptide translation Azithromycin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies azithromycin demonstrates antiparasitic effects by inhibiting parasite propagation with an IC50 value of approximately 8 4 M Based on these pharmacological properties azithromycin holds research potential in investigating protein synthesis mechanisms and studying antimicrobial and antiparasitic drug resistance processes
Catalog No. 50-000-89794 Supplier Apexbio Technology LLC Supplier No. B139825
Azithromycin (CAS 83905-01-5) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to inhibit microbial protein synthesis thereby interfering with peptide translation Azithromycin exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies azithromycin demonstrates antiparasitic effects by inhibiting parasite propagation with an IC50 value of approximately 8 4 M Based on these pharmacological properties azithromycin holds research potential in investigating protein synthesis mechanisms and studying antimicrobial and antiparasitic drug resistance processes