Learn More
Medchemexpress LLC Azithromycin | 83905-01-5 | 98.0% | 748.98 | 1 ML
SDP
Catalog No. 5000387069
Encompass_Preferred
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of a number of bacterial infections. It is intended for research use only.
  • Enhances rhinovirus-induced IFNβ expression in primary bronchial epithelial cells from asthmatics.
  • Represses viral replication.
  • Reduces MMP-9 mRNA and protein levels without affecting NF-κB in endotoxin-challenged monocytic THP-1 cells.
  • Augments expression of interferon-stimulated genes and the pattern recognition receptor MDA5.
Catalog No. 50-003-87069 Supplier Medchemexpress LLC Supplier No. HY1750610MM/1ML
Azithromycin hydrate is a macrolide antibiotic useful for the treatment of a number of bacterial infections. It is intended for research use only.
  • Enhances rhinovirus-induced IFNβ expression in primary bronchial epithelial cells from asthmatics.
  • Represses viral replication.
  • Reduces MMP-9 mRNA and protein levels without affecting NF-κB in endotoxin-challenged monocytic THP-1 cells.
  • Augments expression of interferon-stimulated genes and the pattern recognition receptor MDA5.