Learn More
Medchemexpress LLC Azithromycin-d3 | 163921-65-1 | 98.0% | 752.00 | 1 MG
SDP
Catalog No. 5000386309
Encompass_Preferred
Azithromycin-d3 is the deuterium labeled Azithromycin, a macrolide antibiotic used for the treatment of various bacterial infections. It is suitable for applications requiring a tracer or an internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86309 Supplier Medchemexpress LLC Supplier No. HY17506S1MG
Azithromycin-d3 is the deuterium labeled Azithromycin, a macrolide antibiotic used for the treatment of various bacterial infections. It is suitable for applications requiring a tracer or an internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS