Learn More
Medchemexpress LLC Azithromycin-d3 | 163921-65-1 | 99.7% | 752.00 | 5 MG
SDP
Catalog No. 5000386568
Encompass_Preferred
Azithromycin-d3 is the deuterium labeled Azithromycin. Azithromycin (CP-62993) is a macrolide antibiotic useful for the treatment of a number of bacterial infections. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Azithromycin
  • Useful for treatment of bacterial infections
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development. Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-86568 Supplier Medchemexpress LLC Supplier No. HY17506S5MG
Azithromycin-d3 is the deuterium labeled Azithromycin. Azithromycin (CP-62993) is a macrolide antibiotic useful for the treatment of a number of bacterial infections. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Azithromycin
  • Useful for treatment of bacterial infections
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development. Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.